Dihydrolinalool ≥97.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,7-Dimethyl-6-octen-3-ol, 3,7-Dimethyloct-6-en-3-ol
D1592-25ML
D1592-500ML
TCIAD1592-25MLEA
0
GBP
TCIAD1592-25ML
TCIAD1592-500ML
Dihydrolinalool ≥97.0% (by GC)
Dihydrolinalool
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 200°C Flash Pt: 178°C |
MDL Number:
MFCD00059140 CAS Number: 18479-51-1 EINECS: 242-359-8 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...