1-Chloroethylisopropyl carbonate ≥98.0% (by GC)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Carbonic Acid 1-Chloroethyl Isopropyl Ester, 1-Chloroethyl isopropyl carbonate
C1787-25G
C1787-250G
TCIAC1787-25GEA
0
GBP
TCIAC1787-25G
TCIAC1787-250G
1-Chloroethylisopropyl carbonate ≥98.0% (by GC)
1-Chloroethylisopropyl carbonate
Formula:
C₆H₁₁ClO₃ MW: 166.60 g/mol Boiling Pt: 70°C |
MDL Number:
MFCD04038100 CAS Number: 98298-66-9 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...