Lithium acetate dihydrate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Acetic acid lithium salt

EMR162001 EMR162100 EMR162500
EURLEMR162001EA 0 GBP
EURLEMR162001 EURLEMR162100 EURLEMR162500
Lithium acetate dihydrate
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
MW: 102.02 g/mol
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR