2-Ethoxyethyl acetate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Ethylglycol acetate
M-Clarity™ quality level = MQ200
SIAL109967-2.5LEA
111
GBP
SIAL109967-2.5L
SIAL109967-25ML
SIAL109967-1L
2-Ethoxyethyl acetate, Sigma-Aldrich®
2-Ethoxyethyl acetate
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 156 °C (1013 hPa) Melting Pt: –62 °C Density: 0.976 g/cm³ (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00009251 CAS Number: 111-15-9 EINECS: 203-839-2 UN: 1172 ADR: 3,III Merck Index: 13,03787 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...