2-Ethoxyethyl acetate

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Ethylglycol acetate

PROOCHE163EA 287 GBP
PROOCHE163
2-Ethoxyethyl acetate
2-Ethoxyethyl acetate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.976 g/cm³ (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00009251
CAS Number: 111-15-9
EINECS: 203-839-2
UN: 1172
ADR: 3,III
Merck Index: 13,03787

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR