Ethyl palmitate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Hexadecanoic acid ethyl ester
, Ethyl hexadecanoate
, Hexadecylic acid ethyl ester
, Palmitic acid ethyl ester
EHERC14192000EA
102
GBP
EHERC14192000
Ethyl palmitate
Ethyl palmitate
Formula:
C₁₈H₃₆O₂ MW: 284.48 g/mol Boiling Pt: 342 °C (1013 hPa) Melting Pt: 24 °C Density: 0.86 g/cm³ (20 °C) |
MDL Number:
MFCD00008996 CAS Number: 628-97-7 EINECS: 211-064-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...