L(+)-Lysine monohydrate
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
L-(+)-Lysine, Hydrate
, L-Lysine hydrate
, Lys monohydrate
, H-Lys-OH monohydrate
, (S)-(+)-Lysine monohydrate
, (S)-(+)-2,6-Diaminocaproic acid monohydrate
, K monohydrate
, (S)-(+)-2,6-Diaminohexanoic acid monohydrate
, H-Lys-OH·H₂O
MOLE54686501-100GEA
0
GBP
MOLE54686501-100G
L(+)-Lysine monohydrate
L(+)-Lysine monohydrate
Formula:
H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O MW: 164.2 g/mol Melting Pt: 225 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151035 CAS Number: 39665-12-8 EINECS: 200-294-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...