Yttrium(III) acetylacetonate hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Yttrium (III) tris(2,4-pentanedionato-O,O') hydrateTris(2,4-pentanedionato-O,O')yttrium (III) hydrateYttrium (III) 2,4-pentanedionate hydrate

M-Clarity™ quality level = MQ100

SIAL438790-5GEA 169 GBP
SIAL438790-5G
Yttrium(III) acetylacetonate hydrate, Sigma-Aldrich®
Yttrium (III) acetylacetonate hydrate
Formula: C₁₅H₂₁O₆Y·H₂O
MW: 404.25 g/mol
MDL Number: MFCD00013509
CAS Number: 207801-29-4
EINECS: 239-607-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR