Calcium iodide hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

208477-100G 208477-500G
SIAL208477-100GEA 163 GBP
SIAL208477-100G SIAL208477-500G
Calcium iodide hydrate, Sigma-Aldrich®
Calcium iodide hydrate
Formula: CaI₂·H₂O
MW: 311.9 g/mol
Density: 2.55 g/cm³ (20 °C)
MDL Number: MFCD00149622
CAS Number: 71626-98-7
EINECS: 233-276-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR