Disodium metasilicate nonahydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
SIALS5904-1KGEA
416
GBP
SIALS5904-1KG
Disodium metasilicate nonahydrate, Sigma-Aldrich®
di-Sodium metasilicate nonahydrate
Formula:
Na₂SiO₃·9H₂O MW: 284.2 g/mol Melting Pt: 40 °C Density: 2.62 g/cm³ (21 °C) |
CAS Number:
13517-24-3 EINECS: 229-912-9 UN: 3253 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...