Sodium diethyldithiocarbamate trihydrate

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Diethyldithiocarbamic acid sodium salt trihydrateN,N-Diethyldithiocarbamic acid sodium salt trihydrate(Diethyldithiocarbamato-S,S')sodium trihydrate

MOLE34513940-500GEA 0 GBP
MOLE34513940-500G
Sodium diethyldithiocarbamate trihydrate
Sodium diethyldithiocarbamate trihydrate
Formula: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Melting Pt: 95…98.5 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00150617
CAS Number: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9,III
Merck Index: 12,03443

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR