DL-Lithium lactate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-2-Hydroxypropanoic acid lithium saltDL-Lactic acid lithium salt(±)-Lactic acid lithium saltLithium 2-hydroxypropanoate(±)-2-Hydroxypropionic acid lithium salt

M-Clarity™ quality level = MQ200

SIAL440469-50GEA 98.6 GBP
SIAL440469-50G
DL-Lithium lactate, Sigma-Aldrich®
DL-Lithium lactate
Formula: H₃CCH(OH)COOLi
MW: 96.01 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00065399
CAS Number: 867-55-0
EINECS: 212-761-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR