Lactose monohydrate
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Lactobiose
, Milk sugar
SERA27451.02EA
0
GBP
SERA27451.02
Lactose monohydrate
Lactose monohydrate
Formula:
C₁₂H₂₂O₁₁·1H₂O MW: 360.32 g/mol Melting Pt: 223 °C Storage Temperature: Ambient |
CAS Number:
10039-26-6 EINECS: 200-559-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...