Palladium ≥99.95%, powder - 60 mesh

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

STRE93-4631-1GEA 0 GBP
STRE93-4631-1G
Palladium ≥99.95%, powder - 60 mesh
Palladium
Formula: Pd
MW: 106.420 g/mol
Boiling Pt: 3140 °C
Melting Pt: 1552 °C
Density: 12.02
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR