Methylene blue NZ, TECHNICAL, extra

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

34560.265EA 144 GBP
34560.265
Methylene blue NZ, TECHNICAL, extra
Methylene blue NZ
Formula: C₃₂H₃₆Cl₄N₆S₂Zn
MW: 776.01 g/mol
CAS Number: 6372-69-6
EINECS: 228-905-8

Order Now

Specification Test Results

Identification Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR