Light distillate ≥99.9995% (metals basis) ICP solvent for inorganic trace analysis, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

54362-1LEA 71.5 GBP
54362-1L
Light distillate ≥99.9995% (metals basis) ICP solvent for inorganic trace analysis, Supelco®
Light distillate

ICP solvent is low odor and reduced toxicity equivalent to classical kerosene or xylene and ideal for dilution of oils and other organic fluids for determination of metal traces.

Boiling Pt: 200…250 °C (lit.)
CAS Number: 64742-47-8
UN: 1993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR