4-Aminophenol ≥99.0%, Msynth® plus for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Hydroxyaniline

M-Clarity™ quality level = MQ400

8.45129.1000EA 182 GBP
8.45129.1000
4-Aminophenol ≥99.0%, Msynth® plus for synthesis, Sigma-Aldrich®
4-Aminophenol
Formula: H₂NC₆H₄OH
MW: 109.13 g/mol
Boiling Pt: 284 °C (1013 hPa) (dec.)
Melting Pt: 190 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00007869
CAS Number: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
https://uk.vwr.com/stibo/web/std.lang.all/04/69/2300469.jpg

Order Now

Specification Test Results

Colour (visual) white to off white
Appearance of substance (visual) powder
Assay (HClO₄) ≥ 99.0 %
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR