Organic reference standard, disperse orange 3, analytical standard, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
53882-25MGEA 141 GBP
53882-25MG
Organic reference standard, disperse orange 3, analytical standard, Supelco®
Standards Organic Standards
Used to provide experimental verification of a two-level model for the enhancement of the nonresonant, nonlinear refractive index, with a decrease in absorption by reduction of a donor group strength.