Acetic acid:chloroform (3:2) (v/v), pure

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

APPC145365.1212EA 96.8 GBP
APPC145365.1212
Acetic acid:chloroform (3:2) (v/v), pure
Acetic acid:chloroform
Storage Temperature: Ambient
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Chloroform (A2640) 2 Parts (v/v)
Acetic Acid (A0662) 3 Parts (v/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR