Giemsa's stain improved solution R66, Gurr™ for microscopical staining
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Giemsa's azur-eosin-methylene blue
, Azure mixture sicc. Giemsa stain
, Azur-Eosin-Methylene blue acc. Giemsa
350865PEA
99.7
GBP
350865P
350864X
Giemsa's stain improved solution R66, Gurr™ for microscopical staining
Giemsa's stain
MDL Number:
MFCD00081642 CAS Number: 51811-82-6 |
Specification Test Results
Suitability for microscopy (blood smear) | Passes test |
Erythrocytes | Beige-brownish |
Nuclei | Violet |
Eosinophilic granules | Reddish to red-brown |
Neutrophilic granules | Red-violet |
Lymphocyte cytoplasm | Blue |
Monocyte plasma | Grey-blue |
Thrombocytes | Violet |
Basophilic granules | Dark violet |
Blood parasite nuclei | Light red |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...