Vacuolin-1 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(2-(3-Iodobenzylidene)hydrazinyl)-6-morpholino-N,N-diphenyl-1,3,5-triazin-2-amine3-Iodobenzaldehyde[4-(diphenylamino)-6-(4-morpholinyl)-1,3,5-triazin-2-yl]hydrazone

Packaged under inert gas.

673000-10EA 160 GBP
673000-10
Vacuolin-1 ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Vacuolin-1

A cell-permeable triazine based compound that inhibits Ca²⁺-dependent fusion of lysosomes with the plasma membrane and the release of lysosomal content, without affecting enlargeosomes or other membrane-bound organelles.


It does not disturb the organisation of the actin or tubulin cytoskeletal network, nor does it interfere with membrane traffic along the constitutive pathway from the ER to the plasma membrane.


Caution: Protect from light.

Formula: C₂₆H₂₄IN₇O
MW: 577.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00321410
CAS Number: 351986-85-1

Order Now

Specification Test Results

Purity (by HPLC) ≥95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR