SPE columns, ISOLUTE® PE-AX
Supplier: Biotage
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ISOLUTE®
ARGO503-0100-BEA 0 GBP
ARGO503-0100-B
SPE columns, ISOLUTE® PE-AX
Chromatography Columns
ISOLUTE PE-AX is a silica-based sorbent with a chemically bonded quaternary amine functional group. It is used for the isolation of a wide range of acidic compounds using both catch and release and scavenging SPE protocols.
Nominal capacity of sorbent: 0,6 meq/g.