BCIP/NBT kit
Supplier: BiotiumTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
This kit contains 100 mg each of BCIP sodium salt (10001) and the oxidant NBT (Nitro blue tetrazolium, 10008).
BCIP is the most widely used chromogenic phosphatase substrate, which forms a dark blue precipitate on enzymatic hydrolysis. It is often used with the oxidant NBT (nitro blue tetrazolium chloride), which facilitates the precipitation. BCIP/NBT produces a dark blue/violet precipitate product that can be mounted with permanent or aqueous mounting medium.
Stored desiccated at ≤ 4 °C upon receipt. Protect components from light, especially when in solution.