Fosmidomycin sodium salt ≥95%, crystalline solid
Supplier: Cayman Chemical
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Phosphonic acid, (3-(formylhydroxyamino)propyl), monosodium salt
, Antibiotic FR31564
CAYM16000-5EA
429
GBP
CAYM16000-5
CAYM16000-10
CAYM16000-1
CAYM16000-25
Fosmidomycin sodium salt ≥95%, crystalline solid
Fosmidomycin sodium salt
Fosmidomycin is an antibiotic originally isolated from culture broths of bacteria of the genus Streptomyces that inhibits DOXP reductoisomerase, a key enzyme in the non-mevalonate pathway of isoprenoid biosynthesis. Fosmidomycin is active against both Gram-negative and Gram-positive bacteria and the human malarial parasite P. falciparum (IC50 = 290-370 nM).
Formula:
C₄H₉NO₅P·Na MW: 205.1 g/mol Storage Temperature: Freezer |
CAS Number:
66508-37-0 |
Specification Test Results
Purity | ≥95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...