Preparative HPLC columns, µBondapak®
Supplier: Waters
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WATE027324EA 0 GBP
WATE027324
Preparative HPLC columns, µBondapak®
µBondapak® C18 columns are general purpose, silica-based, reversed phase columns.
  • Suitable for preparative chromatography