Preservative, grotan® OK
Supplier: Schülke
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCUEGROTANOKEA 0 GBP
SCUEGROTANOK
Preservative, grotan® OK
Cleaning Agents Disinfectants
grotan® OK is suitable for use in water-mixed metalworking fluids, metalworking fluid concentrates and other technical emulsions.
  • Bactericidal effect
  • Broad, balanced spectrum of effect (incl. sulphate-reducing bacteria)
  • Immediate effect with improved long term efficacy
  • Demonstrable effective against Mycobacteria
  • Good anticorrosion properties
Contains no nitrate, nitrosing agents or organically bound chlorine (has no effect on the AOX value). Odour improved methylenebisoxazolidine.