Chromatomiser TLC sprayer
Supplier: IBS Stock Items
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IBSA1128309EA 0 GBP
IBSA1128309
Chromatomiser TLC sprayer
Atomisers
Chromatomiser TLC sprayers are based on simple technology where the aerosol compartment is situated above the reservoir.
A small press of the lever lightly pressurises the lower glass reservoir releasing a fine mist of spray. Ideal for TLC and virtually any other spraying applications where an even light film is required - paint formula or sulution.
Delivery information: They are supplied as a pack of 3 complete units with 3 top units and 3 glass reservoirs.