Acetic acid 20%

Supplier: DISTRIBUTOR UK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

DIUKWAC1031LEA 37.4 GBP
DIUKWAC1031L
Acetic acid 20%
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://uk.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR