Blue light transilluminator, Skylight Super-Blue
Supplier: Vilber Lourmat
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Skylight Super-Blue
VILB213120171EA 1380 GBP
VILB213120171
Blue light transilluminator, Skylight Super-Blue
Transilluminators Blue Light Transilluminators
The Skylight Super-Blue transilluminator eliminates the damage caused by UV light on DNA and RNA gels. It also improves cloning efficiency dramatically by eliminating the effects of UV induced nicking or crosslinking, often encountered during the purification of DNA from gels for further use.