2-Nitrothiophene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

N27004-10G N27004-25G
SIALN27004-10GEA 69.2 GBP
SIALN27004-10G SIALN27004-25G
2-Nitrothiophene, Sigma-Aldrich®
2-Nitrothiophene
Formula: C₄H₃NO₂S
MW: 129.14 g/mol
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: 46,5 °C
Density: 1,3644 g/cm³ (42.6 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005425
CAS Number: 609-40-5
EINECS: 210-190-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR