2-Chloro-3-nitrothiophene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF409.2.5 CFF409.5 CFF409.10 005876-5G 005876-1G
CHMPCFF409.2.5EA 0 GBP
CHMPCFF409.2.5 CHMPCFF409.5 CHMPCFF409.10 FLUO005876-5G FLUO005876-1G
2-Chloro-3-nitrothiophene
2-Chloro-3-nitrothiophene
Formula: C₄H₂ClNO₂S
MDL Number: MFCD00052159
CAS Number: 5330-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR