3,4-Lutidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dimethylpyridine

M-Clarity™ quality level = MQ100

L4001-100G L4001-500G
SIALL4001-100GEA 115 GBP
SIALL4001-100G SIALL4001-500G
3,4-Lutidine, Sigma-Aldrich®
3,4-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.955 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006403
CAS Number: 583-58-4
EINECS: 209-511-5
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR