4-Benzylpyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

SIALB30405-5GEA 31.2 GBP
SIALB30405-5G
4-Benzylpyridine, Sigma-Aldrich®
4-Benzylpyridine
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 287 °C (1013 hPa)
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006443
CAS Number: 2116-65-6
EINECS: 218-319-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR