2-Benzoylpyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Phenyl(pyridin-2-yl)methanone, Phenyl 2-pyridyl ketone

M-Clarity™ quality level = MQ100

B14000-100G B14000-25G
SIALB14000-100GEA 133 GBP
SIALB14000-100G SIALB14000-25G
2-Benzoylpyridine, Sigma-Aldrich®
2-Benzoylpyridine
Formula: C₁₂H₉NO
MW: 183.21 g/mol
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 40…43 °C
Density: 1.171 g/cm³ (0 °C)
MDL Number: MFCD00006300
CAS Number: 91-02-1
EINECS: 202-034-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR