3-Amino-4-methylpyridine 98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
L19730.14
L19730.06
L19730.14EA
148
GBP
L19730.14
L19730.06
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine
Purity: 98%
CAS No.: 3430-27-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Odour: Unpleasant
Melting Point: 102-106°
Boiling Point: 254°
Beilstein Reference: 107792
UN No.: 2811
MDL No.: MFCD00128871
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 254 °C Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128871 CAS Number: 3430-27-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...