2-(2-Aminoethyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine

M-Clarity™ quality level = MQ100

A55306-10G A55306-1G A55306-50G
SIALA55306-10GEA 260 GBP
SIALA55306-10G SIALA55306-1G SIALA55306-50G
2-(2-Aminoethyl)pyridine, Sigma-Aldrich®
2-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 49…50 °C (0.03 mmHg)
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR