2-Pyridinethiol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Pyridine-2-thiol, 2-Mercaptopyridine

023265-25G 023265-250G
FLUO023265-25GEA 0 GBP
FLUO023265-25G FLUO023265-250G
2-Pyridinethiol
2-Pyridinethiol
Formula: C₅H₅NS
MW: 111.17 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 132,5…133.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006285
CAS Number: 2637-34-5
EINECS: 220-131-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR