3,4-Diaminopyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine

D7148-5G D7148-1G
SIALD7148-5GEA 91.3 GBP
SIALD7148-5G SIALD7148-1G
3,4-Diaminopyridine, Sigma-Aldrich®
3,4-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 218…219 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00006401
CAS Number: 54-96-6
EINECS: 200-220-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,03006

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR