2-Amino-3-nitropyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Nitropyridin-2-ylamine, 3-Nitropyridin-2-amine, 3-Nitro-2-pyridylamine

M-Clarity™ quality level = MQ100

113514-25G 113514-5G
SIAL113514-25GEA 185 GBP
SIAL113514-25G SIAL113514-5G
2-Amino-3-nitropyridine, Sigma-Aldrich®
2-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 166…168 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006314
CAS Number: 4214-75-9
EINECS: 224-144-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR