2-Iodopyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

558761-1G 558761-5G
SIAL558761-1GEA 70 GBP
SIAL558761-1G SIAL558761-5G
2-Iodopyridine, Sigma-Aldrich®
2-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: 118…120 °C
Density: 1.928 g/cm³ (25 °C)
MDL Number: MFCD00464928
CAS Number: 5029-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR