2-Chloro-5-hydroxypyridine ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Chloro-5-pyridinol, 6-Chloro-3-pyridinol
L17701.06
L17701.03
L17701.06EA
159
GBP
L17701.06
L17701.03
2-Chloro-5-hydroxypyridine ≥97%
2-Chloro-5-hydroxypyridine
Purity: 97%
CAS No.: 41288-96-4
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Melting Point: 152-158°
MDL No.: MFCD00234050
Formula:
C₅H₄ClNO MW: 129.55 g/mol Melting Pt: 152…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234050 CAS Number: 41288-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...