2-Amino-4-chloropyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine
M-Clarity™ quality level = MQ100
676020-25G
676020-5G
SIAL676020-25GEA
179
GBP
SIAL676020-25G
SIAL676020-5G
2-Amino-4-chloropyridine, Sigma-Aldrich®
2-Amino-4-chloropyridine
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol |
MDL Number:
MFCD04113820 CAS Number: 19798-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...