3-Chloro-2,4,5,6-tetrafluoropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Chlorotetrafluoropyridine

003868-5G 003868-25G 003868.100G
FLUO003868-5GEA 0 GBP
FLUO003868-5G FLUO003868-25G FLUO003868.100G
3-Chloro-2,4,5,6-tetrafluoropyridine
3-Chloro-2,4,5,6-tetrafluoropyridine
Formula: C₅ClF₄N
MW: 185.51 g/mol
Boiling Pt: 117…118 °C (1013 hPa)
Density: 1,64 g/cm³ (20 °C)
MDL Number: MFCD00010624
CAS Number: 1735-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR