2-Chloroisonicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

CFF134.2.5 CFF134.5 010413-5G 010413-25G 010413-100G
CHMPCFF134.2.5EA 0 GBP
CHMPCFF134.2.5 CHMPCFF134.5 FLUO010413-5G FLUO010413-25G FLUO010413-100G
2-Chloroisonicotinic acid
2-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR