2-Chloroisonicotinamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF133.2.5 CFF133.5 017099-2.5G 017099-5G 017099-1G
CHMPCFF133.2.5EA 0 GBP
CHMPCFF133.2.5 CHMPCFF133.5 FLUO017099-2.5G FLUO017099-5G FLUO017099-1G
2-Chloroisonicotinamide
2-Chloroisonicotinamide
Formula: C₆H₅ClN₂O
MDL Number: MFCD00221401
CAS Number: 100859-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR