4-Amino-2-bromopyridine 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-4-pyridylamine, 2-Bromopyridin-4-amine

L20007.03 L20007.06
L20007.03EA 28.4 GBP
L20007.03 L20007.06
4-Amino-2-bromopyridine 97%
4-Amino-2-bromopyridine

Purity: 97% CAS No.: 7598-35-8 Molecular Formula: C5H5BrN2 Formula Weight: 173.01 Melting Point: 96-99° Beilstein Reference: 108928

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01646061
CAS Number: 7598-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR