3,6-Dichloropyridazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

009132-25G 009132-100G 009132-500G
FLUO009132-25GEA 0 GBP
FLUO009132-25G FLUO009132-100G FLUO009132-500G
3,6-Dichloropyridazine
3,6-Dichloropyridazine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 70 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR