2-Chloropyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Chloropyrazine

002754-5G 002754-25G
FLUO002754-5GEA 0 GBP
FLUO002754-5G FLUO002754-25G
2-Chloropyrazine
2-Chloropyrazine
Formula: C₄H₃ClN₂
MW: 114.53 g/mol
Boiling Pt: 153 °C (1013 hPa)
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006124
CAS Number: 14508-49-7
EINECS: 238-517-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR