18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6
M-Clarity™ quality level = MQ200
186651-25G
186651-5G
SIAL186651-25GEA
306
GBP
SIAL186651-25G
SIAL186651-5G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane), Sigma-Aldrich®
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Formula:
C₁₂H₂₄O₆ MW: 264.32 g/mol Boiling Pt: 396 °C (1013 hPa) Melting Pt: 37…39 °C Density: 1.237 g/cm³ (25 °C) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005113 CAS Number: 17455-13-9 EINECS: 241-473-5 Merck Index: 13,02630 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...