Sodium dehydracetate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Dehydracetic acid sodium salt, Dehydroacetic acid sodium salt, Sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide, Sodium dehydroacetate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate

M-Clarity™ quality level = MQ100

689521-10KG 689521-40KG
SIAL689521-10KGEA 986 GBP
SIAL689521-10KG SIAL689521-40KG
Sodium dehydracetate, Sigma-Aldrich®
Sodium dehydracetate
Formula: C₈H₇NaO₄
MW: 190.13 g/mol
Melting Pt: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00040583
CAS Number: 4418-26-2
EINECS: 224-580-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR